Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFB6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260997.1 | NP_001247926.1 | 435 | 68 | ||
GenBank mRNA | JU471682.1 | - | 426 | 68 | ||
JV045364.1 | - | 435 | 68 | |||
JV469547.1 | - | 436 | 68 | |||
JV635843.1 | - | 364 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Macaca mulatta |