Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BRIX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261247.1 | NP_001248176.1 | 823 | 65 | ||
GenBank mRNA | JU322347.1 | - | 815 | 65 | ||
JU473284.1 | - | 823 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Macaca mulatta |