Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUDT9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261782.2 | NP_001248711.1 | 766 | 70 | ||
XM_015138743.2 | XP_014994229.1 | 639 | 70 | |||
XM_015138745.2 | XP_014994231.1 | 575 | 70 | |||
GenBank mRNA | JU323329.1 | - | 823 | 70 | ||
JU476026.1 | - | 844 | 70 | |||
JV044417.1 | - | 766 | 70 | |||
JV565138.1 | - | 931 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Macaca mulatta |