Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YBEY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001265798.1 | NP_001252727.1 | 323 | 103 | ||
XM_015132651.2 | XP_014988137.1 | 344 | 103 | |||
XM_015132652.2 | XP_014988138.1 | 669 | 103 | |||
XM_028845073.1 | XP_028700906.1 | 344 | 103 | |||
XM_028845075.1 | XP_028700908.1 | 344 | 103 | |||
GenBank mRNA | JU323926.1 | - | 305 | 103 | ||
JV636225.1 | - | 323 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |