Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001265937.2 | NP_001252866.1 | 1102 | 91 | ||
XM_015143344.2 | XP_014998830.1 | 1220 | 91 | |||
XM_028850494.1 | XP_028706327.1 | 1182 | 91 | |||
GenBank mRNA | JV047233.1 | - | 1100 | 91 | ||
JV635486.1 | - | 1102 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Macaca mulatta |