Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRYM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266891.1 | NP_001253820.1 | 560 | 103 | ||
GenBank mRNA | JU333032.1 | - | 560 | 103 | ||
JV673586.1 | - | 562 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |