Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LXN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266988.1 | NP_001253917.1 | 552 | 71 | ||
GenBank mRNA | JU335536.1 | - | 555 | 71 | ||
JU476463.1 | - | 552 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Macaca mulatta |