Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Drd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012547.1 | NP_036679.1 | 1 - 2 | 313 | 91 | |
XM_006242979.2 | XP_006243041.1 | 1 - 2 | 94 | 91 | ||
GenBank mRNA | M36831.1 | - | 1 - 2 | 94 | 91 | |
X53278.1 | - | 1 - 2 | 93 | 91 | ||
X56065.1 | - | 1 - 2 | 313 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor Rattus norvegicus Probe spans exons |