Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scn3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013119.1 | NP_037251.1 | 1 - 2 | 365 | 91 | |
GenBank mRNA | Y00766.1 | - | 1 - 2 | 365 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Ion Channel Rattus norvegicus Inventoried Probe spans exons |