Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc3a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019283.3 | NP_062156.2 | 1 - 2 | 219 | 63 | |
GenBank mRNA | AB015433.1 | - | 1 - 2 | 185 | 63 | |
CB747485.1 | - | 1 - 2 | 219 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Rattus norvegicus Inventoried Probe spans exons |