Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rfng | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021849.1 | NP_068621.1 | 6 - 7 | 838 | 87 | |
XM_006247910.2 | XP_006247972.1 | 6 - 7 | 904 | 87 | ||
GenBank mRNA | AB016486.1 | - | 6 - 7 | 838 | 87 | |
BC062031.1 | - | 6 - 7 | 883 | 5623611 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Inventoried Probe spans exons |