Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hes3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022687.1 | NP_073178.1 | 1 - 2 | 254 | 74 | |
XM_008764381.1 | XP_008762603.1 | 2 - 3 | 351 | 74 | ||
XM_008764382.1 | XP_008762604.1 | 2 - 3 | 287 | 74 | ||
XM_008764383.1 | XP_008762605.1 | 2 - 3 | 229 | 74 | ||
GenBank mRNA | D13418.1 | - | 1 - 2 | 254 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |