Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prl8a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022846.2 | NP_074037.1 | 2 - 3 | 215 | 92 | |
XM_006253906.1 | XP_006253968.1 | 2 - 3 | 275 | 92 | ||
GenBank mRNA | BC097316.1 | - | 2 - 3 | 216 | 7442457 | 92 |
FQ219955.1 | - | 2 - 3 | 275 | 92 | ||
FQ220067.1 | - | 2 - 3 | 274 | 92 | ||
FQ220110.1 | - | 2 - 3 | 275 | 92 | ||
FQ229702.1 | - | 2 - 3 | 273 | 92 | ||
FQ229765.1 | - | 2 - 3 | 272 | 92 | ||
FQ229882.1 | - | 2 - 3 | 275 | 92 | ||
L06441.1 | - | 2 - 3 | 217 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus 1700 Inventoried Probe spans exons |