Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gnai2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031035.3 | NP_112297.1 | 7 - 8 | 1031 | 89 | |
XM_006243858.1 | XP_006243920.1 | 7 - 8 | 1061 | 89 | ||
GenBank mRNA | AY899210.1 | - | 7 - 8 | 879 | 89 | |
BC078806.1 | - | 7 - 8 | 983 | 7121183 | 89 | |
DQ120471.1 | - | 7 - 8 | 940 | 89 | ||
DQ120472.1 | - | 7 - 8 | 940 | 89 | ||
M12672.1 | - | 7 - 8 | 927 | 89 | ||
M17528.1 | - | 7 - 8 | 1029 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Inventoried Probe spans exons |