Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lum | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031050.1 | NP_112312.1 | 2 - 3 | 909 | 130 | |
GenBank mRNA | BC061878.1 | - | 1 - 2 | 903 | 5599252 | 130 |
FQ221670.1 | - | 2 - 3 | 976 | 130 | ||
FQ229097.1 | - | 2 - 3 | 972 | 130 | ||
FQ230118.1 | - | 1 - 2 | 738 | 130 | ||
X84039.1 | - | 2 - 3 | 909 | 130 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |