Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smcp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031536.2 | NP_113724.1 | 1 - 2 | 151 | 79 | |
GenBank mRNA | BC078795.1 | - | 1 - 2 | 62 | 7134574 | 79 |
U48702.1 | - | 1 - 2 | 157 | 79 | ||
X87883.1 | - | 1 - 2 | 143 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Inventoried Probe spans exons |