Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lxn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031655.1 | NP_113843.1 | 4 - 5 | 678 | 99 | |
GenBank mRNA | BC081763.1 | - | 4 - 5 | 664 | 7125782 | 99 |
FQ220594.1 | - | 4 - 5 | 693 | 99 | ||
FQ229501.1 | - | 4 - 5 | 670 | 99 | ||
U40260.1 | - | 4 - 5 | 645 | 99 | ||
X76985.1 | - | 4 - 5 | 678 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |