Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc18a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031663.2 | NP_113851.1 | 1 | 846 | 99 | |
GenBank mRNA | U09211.1 | - | 1 | 846 | 99 | |
U09838.1 | - | 1 | 103 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Transporter Detect Genomic DNA Rattus norvegicus Inventoried Both primers and probe map within a single exon |