Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ldlr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175762.2 | NP_786938.1 | 1 - 2 | 218 | 91 | |
GenBank mRNA | X13722.1 | - | 1 - 2 | 218 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Rattus norvegicus Inventoried Probe spans exons |