Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc35e1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109107.2 | NP_001102577.2 | 1 - 2 | 441 | 65 | |
GenBank mRNA | BF565650.1 | - | 4 - 5 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Transporter Detect Genomic DNA Rattus norvegicus Probe spans exons |