Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc1a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289941.1 | NP_001276870.1 | 3 - 4 | 665 | 65 | |
NM_001289942.1 | NP_001276871.1 | 4 - 5 | 792 | 65 | ||
NM_019225.2 | NP_062098.1 | 4 - 5 | 782 | 65 | ||
GenBank mRNA | AF265360.1 | - | 3 - 4 | 504 | 65 | |
S59158.1 | - | 4 - 5 | 724 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Rattus norvegicus Probe spans exons |