Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cfi | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024157.1 | NP_077071.1 | 11 - 12 | 1209 | 72 | |
XM_008761526.1 | XP_008759748.1 | 12 - 13 | 1547 | 72 | ||
GenBank mRNA | BC089798.1 | - | 11 - 12 | 1214 | 7321559 | 72 |
Y18965.1 | - | 11 - 12 | 1209 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Rattus norvegicus Probe spans exons |