Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Galnt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024373.1 | NP_077349.1 | 4 - 5 | 789 | 97 | |
XM_006254471.2 | XP_006254533.1 | 5 - 6 | 2210 | 97 | ||
GenBank mRNA | U35890.1 | - | 4 - 5 | 789 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |