Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aco2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024398.2 | NP_077374.2 | 7 - 8 | 943 | 97 | |
GenBank mRNA | AJ243266.1 | - | 7 - 8 | 944 | 97 | |
BC061999.1 | - | 7 - 8 | 943 | 5621226 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |