Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif2ak3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031599.2 | NP_113787.1 | 7 - 8 | 1486 | 63 | |
XM_006236623.1 | XP_006236685.1 | 7 - 8 | 1004 | 63 | ||
XM_006236624.2 | XP_006236686.1 | 7 - 8 | 1524 | 63 | ||
XM_008762977.1 | XP_008761199.1 | 6 - 7 | 1118 | 63 | ||
GenBank mRNA | AF096835.1 | - | 7 - 8 | 1487 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Rattus norvegicus Probe spans exons |