Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031669.1 | NP_113857.1 | 1 - 2 | 91 | 72 | |
GenBank mRNA | AF214733.1 | - | 1 - 2 | 91 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |