Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dusp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053883.2 | NP_446335.1 | 1 - 2 | 832 | 80 | |
GenBank mRNA | AY724528.1 | - | 1 - 2 | 833 | 80 | |
BC087003.1 | - | 1 - 2 | 832 | 7124105 | 80 | |
U42627.1 | - | 1 - 2 | 759 | 80 | ||
X94185.1 | - | 1 - 2 | 625 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Phosphatase Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |