Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grm4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022666.1 | NP_073157.1 | 499 | 85 | ||
GenBank mRNA | M90518.1 | - | 499 | 85 | ||
U47331.1 | - | 499 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor Rattus norvegicus Probe spans exons |