Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Birc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023987.3 | NP_076477.3 | 9 - 10 | 1845 | 89 | |
XM_006242499.2 | XP_006242561.1 | 8 - 9 | 3808 | 89 | ||
XM_006242500.2 | XP_006242562.1 | 9 - 10 | 2578 | 89 | ||
XM_006242502.2 | XP_006242564.1 | 8 - 9 | 2077 | 89 | ||
XM_006242503.2 | XP_006242565.1 | 8 - 9 | 2036 | 89 | ||
GenBank mRNA | AF183430.1 | - | 7 - 8 | 1613 | 89 | |
BC083555.1 | - | 9 - 10 | 1845 | 7112819 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Apoptosis 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |