Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bhmt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030850.1 | NP_110477.1 | 3 - 4 | 359 | 62 | |
GenBank mRNA | AF038870.1 | - | 3 - 4 | 359 | 62 | |
BC078811.1 | - | 3 - 4 | 346 | 7130736 | 62 | |
U96133.1 | - | 3 - 4 | 348 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |