Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snap25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270575.1 | NP_001257504.1 | 2 - 3 | 277 | 75 | |
NM_001270576.1 | NP_001257505.1 | 2 - 3 | 189 | 75 | ||
NM_030991.3 | NP_112253.1 | 2 - 3 | 277 | 75 | ||
GenBank mRNA | AB003991.1 | - | 68 | 75 | ||
AB003992.1 | - | 1 - 2 | 68 | 75 | ||
AF245227.1 | - | 2 - 3 | 276 | 75 | ||
BC087699.1 | - | 2 - 3 | 232 | 7314318 | 75 | |
DY315301.1 | - | 2 - 3 | 209 | 75 | ||
FM033263.1 | - | 2 - 3 | 189 | 75 | ||
FQ212040.1 | - | 2 - 3 | 248 | 75 | ||
FQ213289.1 | - | 2 - 3 | 245 | 75 | ||
FQ213923.1 | - | 2 - 3 | 248 | 75 | ||
FQ214162.1 | - | 2 - 3 | 277 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |