Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scnn1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031548.2 | NP_113736.1 | 6 - 7 | 1697 | 75 | |
GenBank mRNA | BC070902.1 | - | 6 - 7 | 1697 | 7100777 | 75 |
EU627515.1 | - | 6 - 7 | 1332 | 75 | ||
U54699.1 | - | 6 - 7 | 1364 | 75 | ||
U54700.1 | - | 6 - 7 | 1364 | 75 | ||
X70497.1 | - | 6 - 7 | 1410 | 75 | ||
X70521.1 | - | 6 - 7 | 1385 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Ion Channel Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |