Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mybph | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031813.1 | NP_114001.1 | 4 - 5 | 633 | 72 | |
XM_006249888.2 | XP_006249950.1 | 4 - 5 | 743 | 72 | ||
XM_006249889.2 | XP_006249951.1 | 4 - 5 | 742 | 72 | ||
GenBank mRNA | AF077338.1 | - | 4 - 5 | 633 | 72 | |
BC061993.1 | - | 4 - 5 | 672 | 5599482 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |