Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pafah1b3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053654.2 | NP_446106.1 | 3 - 4 | 236 | 63 | |
XM_006228365.2 | XP_006228427.1 | 3 - 4 | 374 | 63 | ||
GenBank mRNA | AF016047.1 | - | 3 - 4 | 236 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |