Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Suclg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053752.2 | NP_446204.2 | 3 - 4 | 336 | 83 | |
GenBank mRNA | BC061537.1 | - | 3 - 4 | 340 | 6920009 | 83 |
FQ212668.1 | - | 3 - 4 | 333 | 83 | ||
FQ214959.1 | - | 3 - 4 | 325 | 83 | ||
FQ215028.1 | - | 3 - 4 | 294 | 83 | ||
FQ215469.1 | - | 3 - 4 | 333 | 83 | ||
FQ216893.1 | - | 3 - 4 | 325 | 83 | ||
FQ217244.1 | - | 3 - 4 | 350 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |