Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Entpd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172030.1 | NP_742027.1 | 2 - 3 | 238 | 59 | |
XM_006233644.2 | XP_006233706.1 | 2 - 3 | 338 | 59 | ||
XM_008761606.1 | XP_008759828.1 | 2 - 3 | 338 | 59 | ||
GenBank mRNA | AF276940.1 | - | 2 - 3 | 238 | 59 | |
BC086558.1 | - | 2 - 3 | 248 | 7302083 | 59 | |
Y11835.1 | - | 2 - 3 | 237 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |