Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dyrk1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012791.2 | NP_036923.1 | 2 - 3 | 131 | 108 | |
XM_006248031.2 | XP_006248093.1 | 2 - 3 | 3076 | 108 | ||
XM_008768562.1 | XP_008766784.1 | 1 - 2 | 2339 | 108 | ||
XM_008768563.1 | XP_008766785.1 | 3 - 4 | 3212 | 108 | ||
XM_008768564.1 | XP_008766786.1 | 2 - 3 | 178 | 108 | ||
XM_008768565.1 | XP_008766787.1 | 2 - 3 | 280 | 108 | ||
GenBank mRNA | X79769.1 | - | 2 - 3 | 133 | 108 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Rattus norvegicus Probe spans exons |