Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mmp11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012980.2 | NP_037112.2 | 2 - 3 | 363 | 71 | |
GenBank mRNA | BC099781.1 | - | 2 - 3 | 357 | 7933545 | 71 |
U46034.1 | - | 2 - 3 | 363 | 71 |
Gene Symbol: LOC103694874 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008773828.1 | XP_008772050.1 | 2 - 3 | 389 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Protease Mammalian Gene Collection Rattus norvegicus Probe spans exons |