Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hcrtr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013074.1 | NP_037206.1 | 2 - 3 | 462 | 91 | |
XM_008766215.1 | XP_008764437.1 | 2 - 3 | 822 | 91 | ||
GenBank mRNA | AF041246.1 | - | 2 - 3 | 462 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor Rattus norvegicus Probe spans exons |