Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001085369.1 | NP_001078838.1 | 18 - 19 | 1930 | 98 | |
NM_134472.3 | NP_608302.2 | 17 - 18 | 1815 | 98 | ||
XM_006238632.2 | XP_006238694.1 | 18 - 19 | 1982 | 98 | ||
XM_006238633.1 | XP_006238695.1 | 17 - 18 | 1746 | 98 | ||
XM_006238634.2 | XP_006238696.1 | 17 - 18 | 2278 | 98 | ||
XM_006238635.2 | XP_006238697.1 | 18 - 19 | 2155 | 98 | ||
XM_006238636.2 | XP_006238698.1 | 18 - 19 | 2151 | 98 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Probe spans exons |