Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270762.1 | NP_001257691.1 | 6 - 7 | 889 | 75 | |
NM_001270763.1 | NP_001257692.1 | 6 - 7 | 895 | 75 | ||
NM_139105.3 | NP_620805.2 | 6 - 7 | 904 | 75 | ||
XM_008759970.1 | XP_008758192.1 | 6 - 7 | 1453 | 75 | ||
GenBank mRNA | AB296102.1 | - | 5 - 6 | 771 | 75 | |
BC070501.1 | - | 6 - 7 | 811 | 7096144 | 75 | |
X62528.1 | - | 6 - 7 | 889 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |