Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc34a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139338.2 | NP_647554.1 | 9 - 10 | 1018 | 72 | |
XM_006233558.2 | XP_006233620.1 | 10 - 11 | 1231 | 72 | ||
XM_006233559.2 | XP_006233621.1 | 10 - 11 | 1347 | 72 | ||
XM_006233560.2 | XP_006233622.1 | 10 - 11 | 1208 | 72 | ||
XM_006233561.2 | XP_006233623.1 | 9 - 10 | 1121 | 72 | ||
XM_006233562.2 | XP_006233624.1 | 9 - 10 | 1237 | 72 | ||
XM_006233564.2 | XP_006233626.1 | 9 - 10 | 1101 | 72 | ||
XM_006233565.2 | XP_006233627.1 | 10 - 11 | 1207 | 72 | ||
XM_006233566.2 | XP_006233628.1 | 9 - 10 | 1213 | 72 | ||
XM_006233567.2 | XP_006233629.1 | 9 - 10 | 1089 | 72 | ||
XM_008761577.1 | XP_008759799.1 | 10 - 11 | 1296 | 72 | ||
GenBank mRNA | AB077042.1 | - | 9 - 10 | 1023 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Detect Genomic DNA Rattus norvegicus Probe spans exons |