Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stx17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145723.1 | NP_663775.1 | 4 - 5 | 544 | 72 | |
XM_006238024.2 | XP_006238086.1 | 5 - 6 | 622 | 72 | ||
GenBank mRNA | AF115435.1 | - | 4 - 5 | 544 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |