Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nr1d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113422.1 | NP_001106893.1 | 5 - 6 | 1458 | 72 | |
NM_145775.2 | NP_665718.2 | 5 - 6 | 1428 | 72 | ||
GenBank mRNA | BC062047.1 | - | 5 - 6 | 1458 | 5598020 | 72 |
DN948099.1 | - | 1 - 2 | 534 | 72 | ||
M25804.1 | - | 5 - 6 | 1428 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Nuclear Receptor Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |