Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scn5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001160162.1 | NP_001153634.1 | 3 - 4 | 593 | 61 | |
NM_013125.2 | NP_037257.1 | 3 - 4 | 593 | 61 | ||
XM_006244076.2 | XP_006244138.1 | 3 - 4 | 763 | 61 | ||
XM_008766670.1 | XP_008764892.1 | 3 - 4 | 1791 | 61 | ||
XM_008766671.1 | XP_008764893.1 | 3 - 4 | 1791 | 61 | ||
GenBank mRNA | AF353637.1 | - | 2 - 3 | 396 | 61 | |
M27902.1 | - | 3 - 4 | 593 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Ion Channel Rattus norvegicus Probe spans exons |