Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc23a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017315.2 | NP_059011.1 | 2 - 3 | 173 | 66 | |
XM_006254602.2 | XP_006254664.1 | 4 - 5 | 554 | 66 | ||
GenBank mRNA | AF080452.1 | - | 2 - 3 | 173 | 66 | |
BC078851.1 | - | 2 - 3 | 173 | 7127602 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |