Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Anxa4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024155.3 | NP_077069.3 | 4 - 5 | 273 | 68 | |
XM_006236841.2 | XP_006236903.1 | 5 - 6 | 360 | 68 | ||
XM_006236842.2 | XP_006236904.1 | 4 - 5 | 896 | 68 | ||
GenBank mRNA | BC085688.1 | - | 4 - 5 | 273 | 7191458 | 68 |
D38224.2 | - | 2 - 3 | 182 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |