Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271058.1 | NP_001257987.1 | 9 - 10 | 1263 | 122 | |
NM_001271059.1 | NP_001257988.1 | 10 - 11 | 1307 | 122 | ||
NM_001271060.1 | NP_001257989.1 | 9 - 10 | 1298 | 122 | ||
NM_001271061.1 | NP_001257990.1 | 9 - 10 | 1232 | 122 | ||
NM_001271062.1 | NP_001257991.1 | 9 - 10 | 1341 | 122 | ||
NM_173096.3 | NP_775119.2 | 9 - 10 | 1421 | 122 | ||
XM_006248146.2 | XP_006248208.1 | 10 - 11 | 1487 | 122 | ||
XM_006248149.2 | XP_006248211.1 | 10 - 11 | 1530 | 122 | ||
XM_008768555.1 | XP_008766777.1 | 9 - 10 | 3423 | 122 | ||
XM_008768556.1 | XP_008766778.1 | 9 - 10 | 3396 | 122 | ||
XM_008768557.1 | XP_008766779.1 | 9 - 10 | 3331 | 122 | ||
GenBank mRNA | BC099784.1 | - | 9 - 10 | 1409 | 7933587 | 122 |
FQ226931.1 | - | 9 - 10 | 1298 | 122 | ||
FQ230542.1 | - | 9 - 10 | 1310 | 122 | ||
FQ231581.1 | - | 10 - 11 | 1375 | 122 | ||
X52711.1 | - | 8 - 9 | 1215 | 122 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |