Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Taar4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175583.1 | NP_783173.1 | 1 | 969 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 G-protein coupled receptor 3' Most Detect Genomic DNA Rattus norvegicus 1700 Both primers and probe map within a single exon |