Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc80 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022543.2 | NP_071988.2 | 5 - 6 | 2706 | 70 | |
GenBank mRNA | AF223677.1 | - | 5 - 6 | 2814 | 70 | |
AY548105.1 | - | 5 - 6 | 2706 | 70 | ||
BC070926.1 | - | 4 - 5 | 2044 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |